1x1 image pixel for data collection
Sabouraud Dextrose Agar, Emmons w/Yeast Extract
Product Image
Thermo Scientific™
Sabouraud Dextrose Agar, Emmons w/Yeast Extract
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Yeast Extract.
Have Questions?
Catalog NumberdimFormat
R01780
Catalog number R01780
Price (USD)
-
Request A Quote
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, Emmons, with Yeast Extract. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20g/L2. Yeast extract is added to the medium as a growth supplement.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Sabouraud Dextrose Agar (Emmons) w/Yeast Extract.
  • Ready-to-use – Convenience of a prepared media.
  • Suitable for isolation of pathogenic fungi from a material containing a large number of other fungi or bacteria.
  • Yeast extract acts as a growth supplement for fastidious fungi.
This medium also contains casein and meat peptones which supply nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Yeast extract is added to the medium as a growth supplement to ensure the growth of fastidious fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Sabouraud, R. 1892. Ann. Dermatol. Syphil. 3:1061-1087.
  2. Emmons, C.W., C.H. Binford, J.P. Utz, and K.J. Kwon-Chung. 1977. Medical Mycology. 3rd ed. Lea and Febiger, Philadelphia, PA.
Specifications
DescriptionSabouraud Dextrose Agar, Emmons w/Yeast Extract
FormatMonoplate Deep Pour
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection