Schaedler Agar w/Blood, Hemin, Vitamin K
Thermo Scientific™
Schaedler Agar w/Blood, Hemin, Vitamin K
Grow and isolate obligate and facultative anaerobic organisms with these prepared plates.
Have Questions?
Catalog number R01800
Price (USD)/ Each
-
Request A Quote
Grow and isolate obligate and facultative anaerobic organisms, such as the fastidious anaerobic bacteria Bacteroides, Prevotella and Porphyromonas, using Thermo Scientific™ Remel™ Schaedler Anaerobe Agar with Sheep Blood, Haemin and Vitamin K1
Schaedler Agar w/Blood, Hemin, Vitamin K contains cysteine hydrochloride and glucose, as reducing substances, with the advantage that cysteine inhibits the growth of Escherichia coli. Schaedler Agar w/Blood, Hemin, Vitamin K has been shown to be a suitable alternative to blood agar for the enumeration of clostridia and has been used for the examination of food, waste products and ditch water. Studies have shown the necessity for strict anaerobic conditions for the successful recovery of obligate anaerobes when using this medium without the addition of blood.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
DescriptionSchaedler Agar w/Blood, Hemin, Vitamin K
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach