Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Opaque light tan |
Description | Spectra UTI Medium |
Format | 85mm Monoplate |
Manufacturing Origin | United States |
Product Type | Agar |
Fill Volume | 18 mL |
Packaging Type | Flow Wrapped - Single Layer |
Quantity | 10/Pk. |
Unit Size | Each |
Color | Opaque light tan |
Description | Spectra UTI Medium |
Format | 85mm Monoplate |
Manufacturing Origin | United States |
Product Type | Agar |
Fill Volume | 18 mL |
Packaging Type | Flow Wrapped - Single Layer |
Quantity | 100/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | dimFormat | Quantity | Manufacturing Origin | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R01818 | Each | Spectra UTI Medium | 10/Pk. | United States | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R01819 | Each | Spectra UTI Medium | 100/Pk. | United States | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|