Spectra™ VRE Medium
Spectra™ VRE Medium
Actual product may vary
Thermo Scientific™
Spectra™ VRE Medium
Obtain fast and accurate results with a chromogenic medium designed for screening clinical samples for the presence of vancomycin-resistant Enterococcus (VRE).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFormatQuantityManufacturing Origin
R01830Spectra VRE Medium85mm Monoplate10/Pk.United States
R01832Spectra VRE Medium85mm Monoplate100/Pk.United States
2 Options
Catalog number R01830
Price (USD)/ Each
-
Request A Quote
Description:
Spectra VRE Medium
Format:
85mm Monoplate
Quantity:
10/Pk.
Manufacturing Origin:
United States
Improve the prevention and control of VRE in healthcare settings by rapidly and reliably isolating and presumptively identifying vancomycin-resistant Enterococcus (VRE) within 24 hours, direct from sample, using Thermo Scientific™ Spectra™ VRE. Vancomycin-resistant Enterococcus faecium and Enterococcus faecalis are differentiated through the utilization of two separate chromogens. Interpretation is aided through the inclusion of a semi-opaque background, providing a distinct visual contrast against light blue and indigo colonies.
Differentiation of vancomycin resistant E. faecium from E. faecalis is achieved through the inclusion of two chromogens that are targeted by specific enzymes: phosphatase and α-galactosidase. The action of these enzymes on the chromogens results in a build-up of color within the colony. The color produced depends on which enzymes the organisms possess. The presence of phosphatase enzymes in both E. faecium and E. faecalis results in a light blue colony; however, E. faecium also produces α-galactosidase resulting in a mix of blue and pink chromophores within the bacterium, producing indigo to purple colonies which are easily distinguished from the light blue E. faecalis colonies.
Additional antibiotics, in combination with vancomycin, are present to suppress the growth of competing flora including E. gallinarum and E. casseliflavus, both of which are intrinsically resistant to vancomycin, possessing the chromosomally encoded VanC resistance mechanism. The VanC resistance mechanism is not readily transmissible between organisms and, as such, is deemed less clinically significant than VanA and VanB mechanisms which are encoded on freely transmissible genetic elements, plasmids and transposons, thus increasing the risk of resistance genes spreading to other organisms.
  • Rapidly and economically detects colonization of Vancomycin-Resistant Enterococcus (VRE) colonization
  • Aids in the prevention and controls the spread of vancomycin resistance in healthcare settings
  • Correctly classifies patients whether positive or negative
  • Provides superior results in a selective and differential chromogenic medium
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
ColorSemi-opaque light tan
DescriptionSpectra VRE Medium
Format85mm Monoplate
Manufacturing OriginUnited States
Product TypeAgar
Fill Volume18 mL
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Unit SizeEach
Contents & Storage
2°C to 8°C