Thermo Scientific™

Yersinia Selective Agar

Catalog number: R01998
Have Questions?
Thermo Scientific™

Yersinia Selective Agar

Catalog number: R01998
Have Questions?
Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples after 18 to 24 hours using Thermo Scientific™ Yersinia Selective Agar (CIN Agar).
 
Catalog Number
R01998
Unit Size
Each
dimFormat
Certifications/Compliance
Industrial Reference: BAM, COMPF.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionYersinia Selective Agar
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Catalog Number
Unit Size
Each
dimFormat
Certifications/Compliance
Industrial Reference: BAM, COMPF.
Price (USD)
Full specifications
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionYersinia Selective Agar
Format20mm x 150mm Pour Tube
Product TypeAgar
Quantity20 x 25 mL
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizedimFormatCertifications/CompliancePrice (USD)
R01998Full specifications
EachIndustrial Reference: BAM, COMPF.Request A Quote
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionYersinia Selective Agar
Format12mm x 85mm Monoplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
R09999Full specifications
EachIndustrial Reference: BAM, COMPF.Request A Quote
Certifications/ComplianceIndustrial Reference: BAM, COMPF.
DescriptionYersinia Selective Agar
Format20mm x 150mm Pour Tube
Product TypeAgar
Quantity20 x 25 mL
Unit SizeEach
Showing 2 of 2
Specifically developed for the optimum growth and recovery of Yersinia enterocolitica, the formulation includes cefsulodin, Irgasan™ and novobiocin. The typical colonies of Yersinia enterocolitica will develop as a red bullseye surrounded by a transparent border and will vary considerably among serotypes in colony size, smoothness and the ratio of the border to center diameter. Most other organisms that are capable of growing will produce larger colonies (>2mm in diameter) with diffuse pinkish centers and opaque outer zones. Serratia liquefaciens, Citrobacter freundii and Enterobacter agglomerans may give a colonial morphology resembling Yersinia enterocolitica. These organisms can be differentiated from Yersinia enterocolitica by biochemical tests.

Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text