Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate
Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate
Thermo Scientific™

Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar Plate

Grow fastidious organisms, detect and enumerate coliforms, and isolate and detect Salmonella and Shigella spp. on a single plate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
R0205010/Pk.85mm biplateTSA-5% Sheep Blood/MacConkeyUnited States
R02049100/Pk.85mm biplateTSA-5% Sheep Blood/MacConkeyUnited States
Catalog number R02050
Price (USD)
-
Request A Quote
Quantity:
10/Pk.
Format:
85mm biplate
Media Type:
TSA-5% Sheep Blood/MacConkey
Manufacturing Origin:
United States
Isolate and cultivate fastidious microorganisms with clearly visible hemolytic reactions (staphylococci and streptococci) and detect and enumerate coliform organisms such as E. coli, while at the same time isolating and detecting Salmonella and Shigella spp., using Thermo Scientific™ Blood Agar (TSA with 5% Sheep Blood) / MacConkey Agar.
Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms while reducing storage capacity issues in the incubator with a bi-plate that includes two widely used microbiological media. One half of the plate contains TSA with 5% sheep blood to promote growth of fastidious organisms with clearly visible hemolysis forms for staphylococci and streptococci. MacConkey Agar on the other half of the plate is a selective medium for differentiating between coliforms and lactose and non-lactose fermenters.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
ColorOpaque red/ clear pink
Format85mm biplate
Manufacturing OriginUnited States
Media TypeTSA-5% Sheep Blood/MacConkey
Product TypeAgar
Fill Volume10 mL
Packaging TypeFlow Wrapped - Single Layer
Quantity10/Pk.
Unit SizeEach
Contents & Storage
2°C to 8°C

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link