Search Thermo Fisher Scientific
Catalog Number | dimFormat |
---|---|
R02060 |
Thermo Scientific™ Remel Columbia CNA w/5% Sheep Blood, EMB Agar, Levine is a biplate used for selective isolation and differentiation of gram-negative enteric and gram-positive bacilli from clinical and industrial samples.
Share catalog number, name or link