Salmonella Shigella/XLD Biplate
Salmonella Shigella/XLD Biplate
Thermo Scientific™
Salmonella Shigella/XLD Biplate
Perform selective and differential isolation of enteric gram-negative bacilli using Salmonella Shigella/XLD Biplate.
Have Questions?
Catalog number R02140
Price (USD)
-
Request A Quote
Perform selective and differential isolation of enteric gram-negative bacilli, especially Salmonella and Shigella, from clinical specimens using Thermo Scientific™ Remel™ Salmonella Shigella(SS)/XLD Biplate. This biplate is used for isolating bacteria such as Salmonella and Shigella spp1-3. Leifson modified the Desoxycholate-Citrate Agar used for the detection of enteric pathogens4 and added bile salts mixture to inhibit the growth of gram-positive organisms. XLD medium was developed by Taylor5.
Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.
  • Ready-to-use – Convenience of a prepared media.
  • Performs screening and isolation on the same dish using two agars.
  • Ease of handling and less product is used.
  • Ease of identification – On SS agar colonies of Salmonella and Shigella are smooth and opaque; some appear to have black centered colonies whereas on XLD agar Salmonella produces red colonies without black center and Shigella colonies are transparent.
  • Selective – Due to the inclusion of selective inhibitory substances brilliant green, bile salts, thiosulphate, and citrate in SS agar and desoxycholate in XLD agar, most gram-positive and coliform organisms are inhibited.
The SS agar contains bile salts, brilliant green and citrates which acts as selective agents and inhibit gram-positive bacteria and coliforms. Sodium thiosulfate, a sulfur source and ferric ammonium citrate work as an indicator to enable organisms which produce H2S to form black-centered colonies, including some strains of Salmonella. Lactose is a differentiator.
The XLD agar contains desoxycholate which acts as a selective agent and inhibits gram-positive organisms. Xylose is fermented by most gram-negative bacilli. Lysine acts as a differentiator for Salmonella spp. from non-pathogenic enteric bacilli. Salmonella produces lysine decarboxylase which causes pH to revert to alkaline after xylose is fermented, producing red colonies. Lactose and sucrose are added in excess to prevent lysine-positive coliforms from reverting to alkaline conditions. Sodium thiosulfate, a sulfur source, and ferric ammonium citrate work as indicators to enable organisms which produce H2S to form black-centered colonies under alkaline conditions.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
  1. Issenberg, H.D., S. Kominos, and M. Siegel. 1969. Appl. Microbiol. 18:656-659.
  2. Issenberg, H.D. 2004. Clinical Microbiology Procedures Handbook. 2nd ASM Press, Washington D.C.
  3. Macfaddin, J.F. 1985. Media for Isolation-Cultivation-Identification-Maintenance of Medical Bacteria. Vol. 1. Williams and Wilkins, Baltimore, MD.
  4. Liefson, E., 1935. J. Pathol. Bacteriol. 40:581.
  5. Taylor, W. I. 1965. Am. J. Clin. Pathol. 44:471-475.
Specifications
DescriptionQC-Slide™ AFB Stain Control; 10/Pk.
FormatBiplate
Product TypeAgar
Quantity10/Pk.
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link