Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:GMP-quality NTPs are available for RNA vaccine and therapeutic development. See details
Thermo Scientific™

NTP Set, 100 mM Solution

Catalog number: R0481
Have Questions?
Thermo Scientific™

NTP Set, 100 mM Solution

Catalog number: R0481
Have Questions?
Catalog Number
R0481
Unit Size
4 x 0.25 mL
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
R04814 x 0.25 mL
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

The Thermo Scientific NTP set is a convenient set of 100 mM solutions of each ATP, CTP, GTP and UTP. These highly stable nucleotides are more than 99% pure and can be used in a variety of molecular biology applications.

Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
in vitro transcription

Related products
100 mM ATP Solution
100 mM CTP Solution
100 mM GTP Solution
100 mM UTP Solution

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
100 mM
For Use With (Application)
in vitro Transcription, RNA Amplification
Shipping Condition
Dry Ice
Conjugate
Unconjugated
Quantity
4 x 0.25 mL
Product Type
NTP Set
Form
Liquid

Contents & Storage

NTP Set, 100 mM solution (4 x 0.25 mL)

Store at –20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text