Thermo Scientific™

TurboFect™ Transfection Reagent

Catalog number: R0534
Have Questions?
Thermo Scientific™

TurboFect™ Transfection Reagent

Catalog number: R0534
Have Questions?
Catalog Number
R0534
Unit Size
5 x 200 µL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 µL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5 x 1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R05345 x 200 µL
-
R0533200 µL
-
R05311 mL
-
R05325 x 1 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific TurboFect Transfection Reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.

Features of TurboFect Transfection Reagent:

• High transfection efficiency of a wide variety of cell types
• No optimization required for most cell lines
• Excellent transfection efficiency in the presence or absence of serum
• Minimal cytotoxicity
• Ready-to-use—no need to reconstitute, dilute or manipulate

Applications of TurboFect Transfection Reagent :
• Stable and transient transfection or co-transfection of DNA or RNA
• Experiments with adherent or suspension cells, differentiated or undifferentiated cells
• Gene expression studies and assays involving GFP or luciferase reporters

TurboFect Transfection Reagent is a sterile solution of a proprietary cationic polymer. TurboFect Reagent interacts with DNA to form small, stable, highly diffusible complexes which are readily endocytosed. The reagent creates a proton-sponge effect that buffers endosomal pH by provoking massive proton accumulation and passive chloride influx. As a result, rapid osmotic swelling of the endocytosed DNA-complex causes it to rupture in the cytosol, allowing translocation of DNA to the nucleus. TurboFect Reagent is ideal for transfection of a variety of cells, including primary and difficult-to-transfect cells.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Type
Transfection Reagent
Sufficient For
500 Transfections
Quantity
200 μL
Transfection Technique
Lipid-Based Transfection
Product Line
TurboFect™

Contents & Storage

Upon receipt store at 4°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text