Water, nuclease-free
Water, nuclease-free
Thermo Scientific™

Water, nuclease-free

Thermo Scientific nuclease-free water is deionized and 0.22 μm membrane-filtered nuclease-free water. It is ideal for all molecular biology applications.Features•Read more
Have Questions?
Change viewbuttonViewtableView
Quantity:
4 x 1.25 mL
30 mL
Catalog number R0581
Price (USD)
20.65
Online Exclusive
26.42
Save 5.77 (22%)
Each
-
Add to cart
Quantity:
4 x 1.25 mL
Request bulk or custom format
Price (USD)
20.65
Online Exclusive
26.42
Save 5.77 (22%)
Each
Add to cart
Thermo Scientific nuclease-free water is deionized and 0.22 μm membrane-filtered nuclease-free water. It is ideal for all molecular biology applications.

Features
• Free of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases
For Research Use Only. Not for use in diagnostic procedures.
Specifications
PackagingVial
Treatment(s)Not DEPC-Treated
GradeMolecular Biology
Quantity4 x 1.25 mL
pH7.6
Unit SizeEach

Frequently asked questions (FAQs)

I would like to order a TaqMan SNP Genotyping Assay. Does the kit include everything I need to perform my experiment or do I need to purchase reagents separately?

Each predesigned TaqMan SNP Genotyping Assay is delivered in a single tube consisting of two differentially labeled allele-specific TaqMan MGB (minor groove binding) probes and a pair of PCR primers. For more information, please see the following link: https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/real-time-pcr-applications/genetic-variation-analysis-using-real-time/snp-genotyping-with-real-time-pcr.html

Besides the TaqMan SNP Genotyping Assay, you would need to purchase a Genotyping Master Mix and nuclease-free water. A list of compatible master mixes can be found in the following link: https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0009593_TaqManSNP_UG.pdf.

Find additional tips, troubleshooting help, and resources within our Gene Expression Analysis & Genotyping Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link