Bile Esculin Agar
Bile Esculin Agar
Actual product may vary
Thermo Scientific™
Bile Esculin Agar
Isolate and differentiate enterococci / Group D streptococci and non-Group D streptococci using this selective microbiological medium.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormatMedia TypeManufacturing Origin
R06029420/Pk.15mm x 103mm Tube - SlantBile Esculin AgarUnited States
R0119010/Pk.MonoplateBile Esculin AgarUnited States
Catalog number R060294
Price (USD)
-
Request A Quote
Quantity:
20/Pk.
Format:
15mm x 103mm Tube - Slant
Media Type:
Bile Esculin Agar
Manufacturing Origin:
United States
Isolate and differentiate enterococci/Group D streptococci and non Group D streptococci using Thermo Scientific™ Bile Esculin Agar, a selective microbiological medium.
Enterococci / Group D streptococci hydrolyse aesculin to form aesculetin and dextrose. Aesculetin combines with ferric citrate in the medium to form a dark brown or black complex which is indicative of a positive result. Bile salts will inhibit Gram-positive bacteria other than enterococci / Group D streptococci.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
Specifications
Format15mm x 103mm Tube - Slant
Media TypeBile Esculin Agar
Quantity20/Pk.
Manufacturing OriginUnited States
Product TypeAgar
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link