Search by lot number or partial lot number
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | 15mm x 103mm Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 5 mL |
Packaging Type | Glass tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | 15mm x 103mm Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 1 mL |
Packaging Type | Glass tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | 15mm x 103mm Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 2 mL |
Packaging Type | Glass tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | 15mm x 103mm Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 2.5 mL |
Packaging Type | Glass tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | 15mm x 103mm Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 4 mL |
Packaging Type | Glass tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | 15mm x 103mm Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 6 mL |
Packaging Type | Glass tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | 16mm x 125mm Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 9.9 mL |
Packaging Type | Glass tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | PB |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 1000 mL |
Packaging Type | Plastic tube with screw cap |
Quantity | Each |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.85% Solution Tubes |
Format | 16mm x 101mm Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 3mL |
Packaging Type | Plastic tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | dimFormat | Quantity | Fill Volume | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R064446 | Each | Saline 0.85% Solution Tubes | 100/Pk. | 5 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R064430 | Each | Saline 0.85% Solution Tubes | 100/Pk. | 1 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R064434 | Each | Saline 0.85% Solution Tubes | 100/Pk. | 2 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R064438 | Each | Saline 0.85% Solution Tubes | 100/Pk. | 2.5 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R064442 | Each | Saline 0.85% Solution Tubes | 100/Pk. | 4 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R064450 | Each | Saline 0.85% Solution Tubes | 100/Pk. | 6 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R07140 | Each | Saline 0.85% Solution Tubes | 100/Pk. | 9.9 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R112560 | Each | Saline 0.85% Solution Tubes | Each | 1000 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R064439 | Each | Saline 0.85% Solution Tubes | 100/Pk. | 3mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|