Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Clear liquid colorless |
Description | Saline 0.45% Solution Tubes |
Format | Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 1.8 mL |
Packaging Type | Glass tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Color | Clear liquid colorless |
Description | Saline 0.45% Solution Tubes |
Format | Tube |
Manufacturing Origin | United States |
Product Type | Buffer Solution |
Fill Volume | 3.0 mL |
Packaging Type | Plastic tube with screw cap |
Quantity | 100/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | dimFormat | Quantity | Fill Volume | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R064462 | Each | Saline 0.45% Solution Tubes | 100/Pk. | 1.8 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R064464 | Each | Saline 0.45% Solution Tubes | 100/Pk. | 3.0 mL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|