Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | Todd Hewitt Broth (LIM Broth) |
Format | 15mm x 103mm Tube |
Manufacturing Origin | United States |
Product Type | Microbiology Media |
Fill Volume | 5 mL |
Quantity | 100/Pk. |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Quantity | Fill Volume | Manufacturing Origin | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R064814 | Each | 100/Pk. | 5 mL | United States | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|