Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

X-Gluc

Catalog number: R0852
Have Questions?
Thermo Scientific™

X-Gluc

Catalog number: R0852
Have Questions?
Catalog Number
R0852
Unit Size
0.5 g
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
0.1 g
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R08520.5 g
-
R08510.1 g
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific X-Gluc (5-bromo-4-chloro-3-indolyl-beta-D-glucuronic acid, cyclohexylammonium salt) is a substrate for beta-glucuronidase (GUS) which is encoded by gusA, a widely used reporter gene. Glucuronidase cleaves X-Gluc to produce colorless glucuronic acid and an intense blue precipitate of chloro-bromoindigo.

Applications

• Detection of GUS expression in plant cells and tissues
• Detection of infections caused by E. coli
• Detection of bacterial contamination in food and water samples.

Note
• Preparation of a 0.1 M stock solution in dimethylformamide or dimethylsulfoxide is recommended.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Colorimetric
Product Type
X-Gluc
Quantity
0.5 g
Substrate Properties
Chemical Substrate
Substrate Type
Beta-Glucuronidase (GUS) Substrate

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text