DTT (dithiothreitol)
Thermo Scientific™

DTT (dithiothreitol)

Thermo Scientific DTT (DL-Dithiothreitol; Clelands reagent) is used to stabilize enzymes and other proteins, which possess free sulfhydryl groups. ItRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R08615 g
R086225 g
Catalog number R0861
Price (USD)
118.65
Online Exclusive
138.00
Save 19.35 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
118.65
Online Exclusive
138.00
Save 19.35 (14%)
Each
Thermo Scientific DTT (DL-Dithiothreitol; Clelands reagent) is used to stabilize enzymes and other proteins, which possess free sulfhydryl groups. It has been shown to restore activity lost by oxidation of these groups in vitro. DTT quantitatively reduces disulfide bonds and maintains monothiols in a reduced state (see Reference 1).

At a final 0.1 M concentration, DTT is also widely used for disruption of protein disulfide bonds in SDS-polyacrylamide gel electrophoresis.

Highlights

• Free of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases.

Applications

• Enzyme stabilization
• Enzymatic reactions
• Protein electrophoresis

Note
• DTT is not stable in solution. Only freshly-made DTT solutions should be used. To prepare a 1 M DTT solution, dissolve 1.55 g of DTT powder in 10 mL of deionized water (e.g. Water, nuclease-free, #R0581).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeDTT (Dithiothreitol)
Quantity5 g
Concentration0.1 M
Unit SizeEach

Frequently asked questions (FAQs)

Can beta-mercaptoethanol (BME) be used rather than DTT as the reducing agent in the NuPAGE LDS Sample Buffer?

Either BME or DTT can be used in the NuPAGE LDS Sample Buffer.

Make sure that a fresh solution of BME is used. FINAL concentration:

DTT 50-100 mM

BME 2-5%

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

How do I prepare a 1 M DTT solution from powder and how should I store the solution?

Preparing 1 M DTT:
1) Dissolve 3.09 g DTT in 20 mL distilled H2O.
2) Dispense into 1 mL aliquots.
3) Store the aliquots at -20 degrees C.
Note: Upon thawing the vial, there may be some precipitation that can be re-dissolved by warming and mixing.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.