Thermo Scientific™

DTT (dithiothreitol)

產品號碼: R0862
Thermo Scientific™

DTT (dithiothreitol)

產品號碼: R0862
產品號碼
R0862
包裝規格
25 g
價格 (TWD)
價格 20,400.00
線上優惠 14,280.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
數量
產品號碼
包裝規格
5 g
價格 (TWD)
價格 4,870.00
線上優惠 3,409.00
(01-Oct-2024 結束)
您的價格
供應狀況
-
數量
產品號碼包裝規格價格 (TWD)供應狀況數量
R086225 g
價格 20,400.00
線上優惠 14,280.00
(01-Oct-2024 結束)
您的價格
-
R08615 g
價格 4,870.00
線上優惠 3,409.00
(01-Oct-2024 結束)
您的價格
-
產品概覽
圖表
影片
建議
建議
文件
常見問題
引用資料與參考文獻
其他資訊
建議
Thermo Scientific DTT (DL-Dithiothreitol; Clelands reagent) is used to stabilize enzymes and other proteins, which possess free sulfhydryl groups. It has been shown to restore activity lost by oxidation of these groups in vitro. DTT quantitatively reduces disulfide bonds and maintains monothiols in a reduced state (see Reference 1).

At a final 0.1 M concentration, DTT is also widely used for disruption of protein disulfide bonds in SDS-polyacrylamide gel electrophoresis.

Highlights

• Free of endo-, exodeoxyribonucleases, ribonucleases, and phosphatases.

Applications

• Enzyme stabilization
• Enzymatic reactions
• Protein electrophoresis

Note
• DTT is not stable in solution. Only freshly-made DTT solutions should be used. To prepare a 1 M DTT solution, dissolve 1.55 g of DTT powder in 10 mL of deionized water (e.g. Water, nuclease-free, #R0581).
For Research Use Only. Not for use in diagnostic procedures.

規格

Concentration
0.1 M
Product Type
DTT (Dithiothreitol)
Quantity
25 g

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text