Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm
<i>Burkholderia cepacia</i> Selective Agar Plated Medium  (USP 60) 60mm
Thermo Scientific™

Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm

Compliance to the USP 60 regulations for B. cepacia complex made simple
Have Questions?
Catalog number R110244
Price (USD)/ Each
-
Request A Quote
Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm is recommended for the qualitative procedures for the selective and differential isolation of B. cepacia complex in purified water systems used in the manufacture of finished products for inhalation use of non-sterile aqueous preparations for oral, oromucosal, cutaneous or nasal use.
Burkholderia cepacia Selective Agar Plated Medium (USP 60) 60mm is recommended for the qualitative procedures of B. cepacia complex for water filtration testing. Confidently detect the presence of B. cepacia in nonsterile medicine with a complete workflow for USP 60 requirements which satisfies all regulations.
  • Compliant formulation according to USP 60
  • Quality tested to meet USP 60 specifically for Pharma customers
  • Part of a complete workflow which include DCM and QCO
For Laboratory Use
For Laboratory Use
Specifications
DescriptionBurkholderia cepacia Sel Agar (USP60) (60mm) 10/PK
Format60 mm petri dish
Product TypeAgar Plate
For Use With (Application)Pharmaceutical purified water testing used in non-sterile manufacturing.
Quantity10/Pk.
Regulatory StatusLab use only
Unit SizeEach