Settling Plate, Sterile, Tryptic Soy Agar (Irradiated)
Settling Plate, Sterile, Tryptic Soy Agar (Irradiated)
Thermo Scientific™
Settling Plate, Sterile, Tryptic Soy Agar (Irradiated)
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Sterile Tryptic Soy Agar (Irradiated).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
R11187010/Pk.Monoplate - Double + Bag
R111872100/Pk.Monoplate - Double + Bag
R11187420/Pk.150mm Plate - Double + Bag
Catalog number R111870
Price (USD)
-
Request A Quote
Quantity:
10/Pk.
Format:
Monoplate - Double + Bag
Ask our AI about this Product
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Sterile Tryptic Soy Agar (Irradiated). It is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. It is also recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
Tryptic Soy Agar is prepared following the standard formula for Soybean-Casein Digest Agar Medium in the United States of Pharmacopeia (USP)5. Tryptic Soy Agar many be supplemented with blood to facilitate the growth of more fastidious bacteria or antimicrobial agents to permit the selection of various microbial groups from mixed flora.
  • Recommended for use in Official Methods of Analysis of AOAC International, IDF, and USDA3,4
  • Ready-to-use – Convenience of prepared medium
This medium contains casein and soy peptones that provide nitrogen, amino acids, and peptides necessary for bacterial growth. Dextrose is a ready source of energy. Sodium chloride supplies essential electrolytes and maintains osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Not intended for IVD use.
  1. Eaton, A.D., L.S. Clesceri, E.W. Rice and A.E. Greenberg. 2005. Standard Methods for the Examination of Water and Wastewater. 21st APHA. Washington, D.C.
  2. Downes, F.P. and K. Ito. 2001. Compendium of Methods for the Microbiological Examination of Foods. 4th APHA, Washington, D.C.
  3. Food and Drug Administration. 2000. Bacteriological Analytical Manual Online. AOAC International, Gaithersburg, MD.
  4. United States Department of Agriculture. 2008. Laboratory Guidebook. Retrieved January 27, 2010.
  5. The United States Pharmacopeia. 2007. 31st ed. United States Pharmacopeial Convention, Rockville, MD.
Specifications
DescriptionSettling Plate Sterile Tryptic Soy Agar
Dimensions (Dia. x H)85 x 13 mm
FormatMonoplate - Double + Bag
Quantity10/Pk.
Product TypeAgar
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?
How does cell culturing on Nunc 384-Well Microplates that have a cell culture-treated surface compare with 96-Well Microplates that have a cell culture-treated surface?
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
281974Certificate of AnalysisJun 25, 2025R111870
277656Certificate of AnalysisJun 16, 2025R111874
277660Certificate of AnalysisJun 16, 2025R111870
265000Certificate of AnalysisJun 14, 2025R111870
276989Certificate of AnalysisJun 14, 2025R111870
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection