Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidin and 2D Barcode.
Color | Ivory, transparent |
Description | Thermo Scientific™ Sterile Triple wrapped Tryptic Soy Agar settling with neutralizers and VHP indicator |
Format | 55 mm plates |
Product Type | Microbiology Media |
Fill Weight | 15 grams (+/- 0.5) |
Packaging Type | No cardboard inner packaging, triple wrapped-irradiated, with VHP barrier wrap |
Quantity | 100/pk |
Shelf Life | 32 weeks |
Unit Size | Each |
Color | Ivory, transparent |
Description | Thermo Scientific™ Sterile Triple wrapped Tryptic Soy Agar settling with VHP indicator |
Format | 85 mm plate |
Product Type | Microbiology Media |
Fill Weight | 25 grams (+/- 0.5) |
Packaging Type | No cardboard inner packaging, triple wrapped-irradiated, with VHP barrier wrap |
Quantity | 100 /pk |
Shelf Life | 32 weeks |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | dimFormat | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R111TW1 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R111TW2 | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Achieve confidence in your results and ensure optimum media growth assurance using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.
Three protective layers reduce risk of contamination:
The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.