Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Format | Quantity |
---|---|---|
R112530 | Serum Bottle, Open Crimp | 10 x 20 mL |
R07770 | Borosilicate Bottle, Septum | 10 x 90 mL |
R112531 | Serum Bottle, Crimp | 10 x 20 mL |
R118610 | 20mm x 113mm Tube | 100 x 9 mL |
Thermo Scientific™ Remel Phosphate Buffer with MgCl2 (pH 7.2) (Buffered Water) APHA Formulation is used as a diluent in microbial limit testing of food and water.
Ordering Information:
REF R112530 has open crimp seal. REF R112531 has complete crimp seal.