Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 200-2,000 cuvette assays using a 2 mL assay volume |
Quantitation Range | 1 to 200 ng |
Product Line | Quant-iT, RIBOGREEN |
Product Type | Quant-iT RiboGreen RNA Reagent |
Shipping Condition | Room Temperature |
Unit Size | 1 mL |
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
Quantitation Range | 3 to 200 ng |
Product Line | RIBOGREEN, RediPlate |
Product Type | RediPlate 96 RiboGreen RNA Kit |
Shipping Condition | Room Temperature |
Unit Size | 1 kit |
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 200-2,000 cuvette assays using a 2 mL assay volume |
Quantitation Range | 1 to 200 ng |
Product Line | Quant-iT, RIBOGREEN |
Product Type | Quant-iT RiboGreen RNA Kit |
Shipping Condition | Room Temperature |
Unit Size | 1 kit |
Catalog Number | Specifications | Unit Size | Product Type | Quantitation Range | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
R11491 | 1 mL | Quant-iT RiboGreen RNA Reagent | 1 to 200 ng | 652.65 Online price 680.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R32700 | 1 kit | RediPlate 96 RiboGreen RNA Kit | 3 to 200 ng | 285.65 Online price 298.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
R11490 | 1 kit | Quant-iT RiboGreen RNA Kit | 1 to 200 ng | 690.65 Online price 718.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|