Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

DNA Loading Dye & SDS Solution (6X)

Catalog number: R1151
Have Questions?
Thermo Scientific™

DNA Loading Dye & SDS Solution (6X)

Catalog number: R1151
Have Questions?
Catalog Number
R1151
Unit Size
5 x 1.0 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R11515 x 1.0 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific 6X DNA Loading Dye & SDS Solution is recommended for agarose gel analysis of DNA samples that contain high amounts of DNA binding proteins. The presence of SDS eliminates DNA-protein interactions and prevents gel-shifts. The high concentration of EDTA protects DNA from degradation by metal-dependent nucleases.

Highlights

• 1% SDS eliminates DNA-protein interactions, prevents appearance of additional bands due to annealing of DNA molecules with cohesive ends
• 100 mM EDTA inhibits metal-dependent nucleases

Applications

• Analysis of DNA samples with high amounts of DNA binding proteins
• Kinetic experiments
• DNA agarose gel analysis after DNA restriction digestions, ligation, or dephosphorylation
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
6X
Gel Compatibility
Acrylamide Gels, Agarose Gels
Label or Dye
Bromophenol Blue, Xylene Cyanol FF (XCFF)
Product Type
DNA Loading Dye & SDS Solution
Quantity
5 x 1 mL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text