Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

IPTG Solution, ready-to-use

Catalog number: R1171
Have Questions?
Thermo Scientific™

IPTG Solution, ready-to-use

Catalog number: R1171
Have Questions?
Catalog Number
R1171
Unit Size
10 x 1.5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R117110 x 1.5 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific IPTG (isopropyl-beta-D-thiogalactopyranoside) is a highly stable synthetic analog of lactose. It inactivates the lac repressor and induces synthesis of beta-galactosidase, an enzyme that promotes lactose utilization. IPTG is used to induce expression of cloned genes under control of the lac operon. It is used in conjunction with X-Gal (#R0941) to determine the lac phenotype in blue/white colony screening.

Applications

• Blue/white colony screening to distinguish recombinant (white) from non-recombinant (blue) colonies
• Induction of gene expression under control of the lac promoter

Note
• For blue/white colony screening use 0.1 mM final IPTG concentration in LB (Luria Broth) media
• Store IPTG Solution at -20°C.

Related Products
IPTG, dioxane-free
IPTG Solution, ready-to-use
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Name or Material
Isopropylthiogalactoside
Concentration
0.1 M
For Use With (Application)
Protein Expression
Product Type
IPTG Solution
Quantity
10 x 1.5 mL
Form
Liquid

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text