A.C.T.™ II Culture Transport System
A.C.T.™ II Culture Transport System
Thermo Scientific™

A.C.T.™ II Culture Transport System

An aerobic, facultative and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.
Have Questions?
Catalog NumberQuantity
R1240210/Pk.
Catalog number R12402
Price (USD)
-
Request A Quote
Quantity:
10/Pk.
Ask our AI about this Product

Thermo Scientific™ Remel A.C.T. II is an aerobic, facultative, and anaerobic culture transport with semi-solid, non-nutritive medium for use with swab or fluid samples.

  • Fluids may be introduced through septum in cap
  • Sterile packaged in individual Tyvek™ envelope with single plastic-shaft rayon-tipped swab for clean environments
Do not freeze or overheat. Do not incubate prior to use.
For in vitro diagnostic use.
Specifications
DescriptionSingle plastic-shaft; 16 x 125mm Tube; 10/Pk.
For Use With (Application)Healthcare
Quantity10/Pk.
Shaft MaterialPlastic
Storage RequirementsStore in original container at 15°C to 30°C until used.
Tip MaterialRayon
Tube Size16 x 125 mm
SterilitySterile
Swab ConfigurationSingle Applicator
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
001656Certificate of AnalysisMay 24, 2024R12402
001657Certificate of AnalysisMay 24, 2024R12402
001659Certificate of AnalysisMay 24, 2024R12402
001658Certificate of AnalysisMay 24, 2024R12402
810256Certificate of AnalysisMay 09, 2024R12402
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection