MicroTest™ M4RT 3mL Tube, Red
MicroTest™ M4RT 3mL Tube, Red
Thermo Scientific™

MicroTest™ M4RT 3mL Tube, Red

Maintain viability of viruses chlamydiae, mycoplasma and urealasma with top quality transport and storage media that comes in four versatile formulations.
Have Questions?
Catalog NumberQuantity
R1250572 Tubes/Pk.
Catalog number R12505
Price (EUR)
-
Request A Quote
Quantity:
72 Tubes/Pk.
Ask our AI about this Product

Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma with top quality media from the experts in media. Thermo Scientific™ MicroTest™ Tubes come in four media formulations to offer flexibility in both transport and storage.

Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae.

MicroTest Tubes come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads.

Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

Trusted reliable results
  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics
Compatibility and viability
  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
Convenient, accommodating format
  • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility
Safe, sturdy packaging
  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
  • Kits include leak-resistant polypropylene bag for safe sample transport

Not all products are available for sale in all territories.

Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Closure ColorRed
CompatibilityCulture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology
DescriptionM4RT 3mL Tube, Red
For Use With (Application)Chlamydiae Viruses
PackagingPackaged in partitioned boxes.
Quantity72 Tubes/Pk.
Shelf Life18 Months
Storage Requirements2°C to 30°C
Transport Temperature2° to 30°C
Volume (Metric)3 mL
FormatTube
Product TypeM4 RT Tube Kit
Unit SizeEach

Intended use statement 

Thermo Scientific™ MicroTest™ M4RT Kit is intended for the collection, transportation, maintenance and storage of viruses and Chlamydiae.

Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
271571Certificate of AnalysisJun 20, 2025R12505
277016Certificate of AnalysisJun 18, 2025R12505
279811Certificate of AnalysisJun 18, 2025R12505
264221Certificate of AnalysisJun 14, 2025R12505
260483Certificate of AnalysisJun 14, 2025R12505
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection