MicroTest™ Tubes
MicroTest™ Tubes
Thermo Scientific™

MicroTest™ Tubes

Maintain viability of viruses chlamydiae, mycoplasma and urealasma with top quality transport and storage media that comes in four versatile formulations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure ColorMedia TypeIncludesQuantity
R12533RedM6, 3 mLGlass Beads72 Tubes/Pk.
R12506RedM4-RTGlass Beads12 Tubes/Pk.
R12515GreenM5Glass Beads72 Tubes/Pk.
R12530RedM6, 1.5 mLGlass Beads72 Tubes/Pk.
Catalog number R12533
Price (USD)
-
Request A Quote
Closure Color:
Red
Media Type:
M6, 3 mL
Includes:
Glass Beads
Quantity:
72 Tubes/Pk.
Maintain organism viability and counts for verification studies, quality control procedures, and repeat testing of viruses, chlamydiae, mycoplasma and ureaplasma with top quality media from the experts in media. Thermo Scientific™ MicroTest™ Tubes come in four media formulations to offer flexibility in both transport and storage.
Designed for the collection, transportation, maintenance and long-term storage of viral specimens, chlamydiae, mycoplasma and ureaplasma, Thermo Scientific MicroTest products are top quality. From formulation to packaging, MicroTest products maintain the viability of organisms through freeze-thaw cycles while inhibiting antimicrobial contaminants and promoting safety.

MicroTest™ M4™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest™ M4RT™ contains gelatin, gentamicin, and amphotericin B for the transport of viruses and Chlamydiae. MicroTest™ M5™ contains vancomycin, amphotericin B, and colistin and protein stabilizers for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest™ M6™ contains gelatin, vancomycin, amphotericin B, and colistin for the transport of viruses, Chlamydiae, Ureaplasmas, and Mycoplasmas.

MicroTest Tubes come with 1.5mL or 3mL of liquid medium in a 15mL conical tube with 3 glass beads.

Keep your lab moving in the right direction with a dependable set of solutions for transport, holding, maintenance and long-term culture storage with the MicroTest family of collection and transport media.

Trusted reliable results
  • Stringent quality control, including cellular toxicity on each lot number
  • Patented formulations with stabilizing proteins, buffers, and specific antibiotics
Compatibility and viability
  • Provides compatible transport for cell culture, enzyme immunoassays, and molecular diagnostic tests
  • Maintains organism viability and counts for better, more reliable results during validation/verification studies, quality control procedures, and repeat testing
Convenient, accommodating format
  • 1.5mL and 3.0mL fill volumes allow multiple tests from one specimen
  • Individual transport media tubes or complete kits offer flexibility and versatility
Safe, sturdy packaging
  • Tough, break-resistant tubes with secure, color-coded screw caps prevent leaking and exposure to infectious agents
  • Kits include leak-resistant polypropylene bag for safe sample transport

Not all products are available for sale in all territories.

Please inquire. Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Closure ColorRed
CompatibilityCulture, Shell vial, Freeze-thaw cycles, Culture maintenance, ELISA, PCR, DNA probe technology
DescriptionM6 Tube
For Use With (Application)Viruses: Chlamydiae, Ureaplasmas, and Mycoplasmas
IncludesGlass Beads
PackagingPackaged in partitioned boxes.
Quantity72 Tubes/Pk.
Shelf Life18 Months
Storage Requirements2°C to 30°C
Transport Temperature2° to 30°C
Volume (Metric)3 mL
FormatTube
Media TypeM6, 3 mL
Product TypeTransport Tube
Unit SizeEach

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link