Thermo Scientific ROX dye is added to the qPCR reactions to serve as an internal reference for normalization of theRead more
Have Questions?
Catalog Number
Quantity
R1371
1 mL
Catalog number R1371
Price (USD)
136.00
Each
In stock
Add to cart
Quantity:
1 mL
Recurring order eligible. Learn more »
Price (USD)
136.00
Each
Add to cart
Ask our AI about this Product
Thermo Scientific ROX dye is added to the qPCR reactions to serve as an internal reference for normalization of the fluorescent signal when using real-time instruments that can detect ROX. ROX allows for the correction of well-to-well variation appearing due to pipetting inaccuracies and fluorescence fluctuations. It can be added to an entire 2X master mix tube or to an individual reaction mixture (for recommended amounts of ROX Solution for a specific real-time instrument refer to the qPCR manual).
ROX does not participate in PCR and does not interfere with the specificity or sensitivity of qPCR assays.
ROX Solution contains ROX dye as a 50 μM aqueous solution at pH 7.6. The excitation maximum and emission maximum of ROX are at 580 nm and 621 nm, respectively.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration50 μM
Fidelity (vs. Taq)2 X
FormatTube
Product LineROX
Product TypeqPCR Reagents
Quantity1 mL
Volume1 mL
Detection MethodPrimer-probe
GC-Rich PCR PerformanceLow
Label or DyeROX
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Contains: • 1 mL ROX Solution, 50 μM
Store at -20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our
Privacy Notice.
Frequently asked questions (FAQs)
Is ROX passive reference dye required for fluorescent signal normalization with the PikoReal instrument?
No, neither ROX nor any other passive reference dye is required when using PikoReal. There is no disadvantage if it is present in the reaction mix, but the software will not use their fluorescence data for normalization of the fluorescence signal of the reporter dyes.