PrimeStore Molecular Transport Medium, 12 mL
PrimeStore Molecular Transport Medium, 12 mL
Thermo Scientific™

PrimeStore Molecular Transport Medium, 12 mL

Non-hazardous, inactivation media for viral collection and transport with Inactivation media for nucleic acid testing and molecular diagnostics.
Have Questions?
Catalog NumberQuantity
R13907100/Pk.
Catalog number R13907
Price (USD)
-
Quantity:
100/Pk.

Stabilize, inactivate and transport infectious unprocessed nasal washes, nasopharyngeal, oral / throat swabs suspected of containing influenza A virus RNA with PrimeStore Molecular Transport Medium® (PS-MTM).

PrimeStore Molecular Transport Medium® is also intended for the stabilization, transportation, and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples.

Specimen collection and transport is a key component in the molecular detection of influenza A and MTB. PS-MTM is a self-contained, ‘ready-to-use’ system that allows for the stabilization and safe transport of clinical samples at ambient temperature from the collection site to the laboratory.

Specifications
ColorClear
CompatibilityThe PS-MTM system is a collection, preservation, transport, and storage system for influenza RNA and MTB DNA. Extraction and purification of nucleic acids have been validated on several manual spin column kits (PrimeXtract™, RNAqueous Micro Kit, Viral RNA Mini Kit, QiaAMP DNA Mini Kit) and automated magnetic bead extraction kits (NucliSENS EasyMAG and MagNA Pure 96 System using the DNA Bacterial/Viral small volume kit). The user is responsible for validating additional extraction and purification kits and platforms.
Fill Volume3 mL
For Use With (Application)Intended for stabilization, transportation, and inactivation of infectious unprocessed nasal washes, nasopharyngeal, oral/throat swabs suspected of containing influenza A virus RNA. PS-MTM is also intended for the stabilization, transportation and inactivation of infectious unprocessed sputum samples suspected of containing Mycobacterium tuberculosis (MTB) DNA from human samples.
Quantity100/Pk.
Shelf Life24 Months
Storage RequirementsRoom temperature, 2°C to 25°C
Tube Size12 mL
Unit SizePack of 100

Save 51% on various cell culture plasticware

Please enter sku to load default questions

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link