Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:GMP-quality NTPs are available for RNA vaccine and therapeutic development. See details
Thermo Scientific™

CTP Solution, Tris buffered

Catalog number: R1451
Have Questions?
Thermo Scientific™

CTP Solution, Tris buffered

Catalog number: R1451
Have Questions?
Catalog Number
R1451
Unit Size
0.25 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R14510.25 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific CTP (cytidine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with Tris base. The nucleotide has greater than 99% purity and is free of nuclease activities and functionally tested in in vitro transcription.

Features
• Purity confirmed by HPLC
• Free of trace contaminating nucleotides, endo-, exodeoxyribonuclease and ribonuclease activities
• Higher in vitro transcription yields using Tris buffered NTPs in comparison to NTPs titrated with NaOH
• Stable for 3 years at -20°C and after multiple freeze-thaw cycles

Applications
• in vitro transcription
• RNA amplification
• siRNA synthesis
• aRNA synthesis

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Shipping Condition
Dry Ice
Concentration
100 mM
Label or Dye
Unlabeled
Quantity
0.25 mL
For Use With (Application)
in vitro Transcription, RNA Amplification
Form
Liquid

Contents & Storage

100 mM CTP, Tris buffered (0.25 mL)

Store at –15 to –25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text