GTP Solution, Tris buffered
GTP Solution, Tris buffered
Thermo Scientific™

GTP Solution, Tris buffered

Thermo Scientific GTP (guanosine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated toRead more
Have Questions?
Catalog NumberQuantity
R14610.25 mL
Catalog number R1461
Price (USD)
53.65
Online Exclusive
61.00
Save 7.35 (12%)
Each
Add to cart
Quantity:
0.25 mL
Request bulk or custom format
Price (USD)
53.65
Online Exclusive
61.00
Save 7.35 (12%)
Each
Add to cart

Thermo Scientific GTP (guanosine 5'-triphosphate), Tris buffered is an extremely stable nucleotide, supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with Tris base. The nucleotide has greater than 99% purity and is free of nuclease activities and functionally tested in in vitro transcription.

Features
• Purity confirmed by HPLC
• Free of trace contaminating nucleotides, endo-, exodeoxyribonuclease and ribonuclease activities
• Higher in vitro transcription yields with Tris buffered NTPs when compared to NTPs titrated with NaOH
• Stable for 3 years at -20°C, and after multiple freeze-thaw cycles

Applications
in vitro transcription
• RNA amplification
• siRNA synthesis
• aRNA synthesis

Related products
ATP, Tris Buffered
UTP, Tris Buffered
NTP Set, Tris Buffered

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)in vitro Transcription, RNA Amplification
FormLiquid
Label or DyeUnlabeled
Quantity0.25 mL
Shipping ConditionDry Ice
Concentration100 mM
Unit SizeEach
Contents & Storage
GTP Solution, Tris buffered, 100 mM (0.25 mL)

Store at –20°C.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link