BactiDisks™ Streptococcus sanguinis Type 1 ATCC™ 10556™
This product is unavailable. For information on equivalent strain with Thermo Scientific™ Culti-Loops™ product portfolio, please contact your local Thermo Scientific representative or customer service department.
BactiDisks™ Streptococcus sanguinis Type 1 ATCC™ 10556™
Thermo Scientific™
BactiDisks™ Streptococcus sanguinis Type 1 ATCC™ 10556™
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
Have Questions?
Catalog NumberQuantity
R1917410 Disk/Vial
Catalog number R19174
Price (USD)
-
Request A Quote
Quantity:
10 Disk/Vial
Ask our AI about this Product
Thermo Scientific™ BactiDisks™ are lyophilized mixtures of charcoal, gelatin and microorganisms in a stable disk format.
The disk is dissolved in 0.1mL of appropriate culture broth, inoculated onto an appropriate culture medium, and incubated at the appropriate temperature, atmosphere and time. BactiDisks are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures. Packaged 10 disks per vial.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.
Specifications
ATCC®10556TM*
DescriptionStreptococcus sanguinis Type 1 ATCC™ 10556™
FormatBactiDisks
Product TypeBactidisk
Target Organism ClassCocci
Test TypeQualitative QC
Quantity10 Disk/Vial
Strain DesignationsWDCM00131, DSS-10
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
977730Certificate of AnalysisApr 15, 2024R19174
915049Certificate of AnalysisApr 15, 2024R19174
900398Certificate of AnalysisApr 15, 2024R19174
865660Certificate of AnalysisApr 15, 2024R19174
857172Certificate of AnalysisApr 15, 2024R19174
5 results displayed, search above for a specific certificate
Product Information
Share catalog number, name or link
1x1 image pixel for data collection