Detect clumping factor and protein A associated with the identification of Staphylococcus aureus while distinguishing S. aureus from other staphylococci.
Have Questions?
Change view
Catalog Number
Description
Quantity
Content And Storage
R21143
BactiStaph Latex Agglutination Test Kit
450 Tests/Kit
R21144
BactiStaph Latex Agglutination Test Kit
150 Tests/Kit
Catalog number R21143
Price (USD)/ Each
-
Request A Quote
Description:
BactiStaph Latex Agglutination Test Kit
Quantity:
450 Tests/Kit
Rapidly identify Staphylococcus aureus and distinguish from other staphylococci by detecting clumping factor and protein A using the Thermo Scientific™ BactiStaph™ Latex Agglutination Test Kit.
Use this rapid test utilizing latex particles which are capable of detecting both clumping factor and protein A simultaneously. Agglutination of white latex particles is visible to the unaided eye within 60 seconds and represents a positive reaction.
Use with suspected S. aureus colonies. Prior to testing, suspect colonies should be Gram stained and a catalase test performed to verify that they are Staphylococcus.
Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Control SetsPositive,Negative
DescriptionBactiStaph Latex Agglutination Test Kit