1x1 image pixel for data collection
Cary-Blair with Indicator Single Vial
Cary-Blair with Indicator Single Vial
Thermo Scientific™
Cary-Blair with Indicator Single Vial
Collect, preserve, and recover enteric bacterial pathogens with a fecal transport that comes with industry color-coding and sporked caps and is stored at room temperature.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberClosure ColorQuantity
R21610Green12 Vials/Pk.
R21925Green120 Vials/Pk.
R21617Green288 Vials/Pk.
R31610Orange12 Vials/Pk.
R31925Orange120 Vials/Pk.
Catalog number R21610
Price (USD)
-
Request A Quote
Closure Color:
Green
Quantity:
12 Vials/Pk.
Ask our AI about this Product
Collect, preserve, and recover enteric bacterial pathogens in stool specimens with Thermo Scientfic™ Cary-Blair with Indicator Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
Maintain the viability of enteric bacterial pathogens during transport and storage with a formulation that includes 1.16% agar and is ready for use without any special preparation. Save valuable storage space with a transport that is stored in its original container at room temperature.
  • Cary-Blair with Indicator is an isotonic and nonnutritive medium, which does not contain a fermentable carbohydrate.
  • Sodium thioglycollate is added to impede oxidation.
  • Disodium phosphate is incorporated in the medium to act as a buffering agent, which prevents overgrowth of Escherichia coli and Enterobacter species.
  • The relatively high pH minimizes the destruction of bacteria due to acid formation.
  • Phenol red, a pH indicator, is added to demonstrate acidic shifts that can affect the viability of some enteric pathogens.
Add convenience and safety to your fecal collection procedures with a broad line of transport products which have a long shelf life and are designed especially for enteric diagnostic testing.
Look to us for convenient kit packaging options, including bulk kits.
Multilingual patient collection instructions are included.
Choose from a wide range of collection and preservation products for fecal specimens, which is important to the successful growth, isolation and detection of microorganisms.
  • Cary Blair with Indicator
  • Formalin 10%
  • Formalin 5%
  • PVA Modified Fixative
  • SAF Fixative
  • Zinc PVA Fixative
  • Empty Vial
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Closure ColorGreen
DescriptionCary-Blair with Indicator Single Vial (Green Cap)
Fill Volume15 mL
Media TypeCary-Blair
PackagingSingle vial. Packaged in partitioned boxes.
Quantity12 Vials/Pk.
Product TypeFecal Transport Vial
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
268872Certificate of AnalysisJul 15, 2025R21925
268480Certificate of AnalysisJul 11, 2025R21610
266548Certificate of AnalysisJul 10, 2025R21617
268541Certificate of AnalysisJul 09, 2025R21617
269438Certificate of AnalysisJul 08, 2025R21610
5 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection