Buffered Glycerol Saline
Buffered Glycerol Saline
Thermo Scientific™
Buffered Glycerol Saline
Maintain viability of enteric pathogens while transporting stool specimens to the lab with a transport medium especially suited for Salmonella and Shigella.
Have Questions?
Catalog number R21650
Price (USD)/ Each
-
Request A Quote
Description:
Buffered Glycerol Saline
Packaging:
Single vial. Packaged in partitioned boxes.
Quantity:
12 Vials/Pk.
Maintain viability of enteric pathogens while transporting stool specimens to the laboratory with Thermo Scientfic™ Buffered Glycerol Saline. Collect, transport and preserve clinical specimens with a transport medium especially suited for Salmonella and Shigella species.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Product TypeBuffered Glycerol Saline
DescriptionBuffered Glycerol Saline
PackagingSingle vial. Packaged in partitioned boxes.
Quantity12 Vials/Pk.
Unit SizeEach