ProSpecT™ Cryptosporidium Microplate Assay
ProSpecT™ Cryptosporidium Microplate Assay
Thermo Scientific™
ProSpecT™ Cryptosporidium Microplate Assay
Rapidly detect Crytosporidium specific antigen from human fecal samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantityContent And Storage
R2454024ProSpecT Cryptosporidium Microplate Assay24 Wells/Kit
R2454096ProSpecT Cryptosporidium Microplate Assay96 Wells/Kit
Catalog number R2454024
Price (USD)
-
Request A Quote
Description:
ProSpecT Cryptosporidium Microplate Assay
Quantity:
24 Wells/Kit
Rapidly detect Crytosporidium specific antigens (CSA) in fresh or preserved fecal specimens using Thermo Scientific™ ProSpecT™ Cryptosporidium Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Crytosporidium. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Cryptosporidium is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures, and common reagents, maximizing operational efficiency, productivity and throughput.
ProSpecT Cryptosporidium provides same-day antigen detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionProSpecT Cryptosporidium Microplate Assay
IncludesTransfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity24 Wells/Kit
Detectable AnalytesCryptosporidium Antigen
Unit SizeEach
Contents & Storage
2°C to 8°C
Intended use statement
A direct, qualitative enzyme immunoassay for the detection of Cryptosporidium specific antigen (CSA) in human faecal samples. Used in a diagnostic workflow to aid clinicians in the diagnosis of Cryptosporidiosis. The device is not automated, is for professional use only and is not a companion diagnostic.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection