Xpect™ Flu A & B Test
Xpect™ Flu A & B Test
Thermo Scientific™
Xpect™ Flu A & B Test
Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens, and get results in 15 minutes.
Have Questions?
Catalog NumberQuantity
R2460020 Tests
Catalog number R24600
Price (USD)
-
Request A Quote
Quantity:
20 Tests
Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens using Thermo Scientific™ Xpect™ Flu A & B Test. Maintain efficiency and productivity with a simple two-step procedure that offers accurate results in 15 minutes. When you need STAT testing or 24/7 coverage, choose Xpect.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Influenza A and Influenza B viral antigens. Reduce secondary testing, decrease unnecessary antibiotic use and guide appropriate antiviral therapy.
Accurate, Easy-to-Read Results
  • Improve patient management with accurate, easy-to-read results in just 15 minutes
  • Eliminate workflow bottlenecks with less than one minute hands-on time and a simple, walk-away procedure
Flexible Options
  • Flexible specimen storage, collection and transport options
  • Recent FDA clearance for analytical detection of cultured influenza A H7N9 virus
Xpect Flu A&B incorporates separate membrane strips for Influenza A and Influenza B. The patient specimen is diluted and added to both sample wells of the device. Antibody-coated color particles bind with target antigen in the diluted sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionXpect Flu A and B Test
IncludesTest devices, specimen diluent (20mL), dilution tubes, disposable transfer pipettes, flu A+/B- control swab and flu B+/A- control swab
Quantity20 Tests
Sample TypeNasopharyngeal
TypeReagent
Detectable AnalytesInfluenza
Unit SizeEach
Contents & Storage
2°C to 8°C
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link