Xpect™ RSV Test
Xpect™ RSV Test
Xpect™ RSV Test
Xpect™ RSV Test
Thermo Scientific™
Xpect™ RSV Test
Rapidly detect Respiratory Syncytial Virus (RSV) from nasopharyngeal specimens in neonatal and pediatric patients and get results in 15 minutes.
Have Questions?
Catalog NumberQuantity
R2460130 Tests
Catalog number R24601
Price (USD)
-
Request A Quote
Quantity:
30 Tests
Ask our AI about this Product
Optimize your workflow by rapidly detecting Respiratory Syncytial Virus antigen (RSV) directly from nasopharyngeal specimens in neonatal and pediatric patients using Thermo Scientific™ Xpect™ RSV Test. Combine excellent clinical performance with a simple, 2-step procedure that offers accurate results within 15 minutes.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Respiratory Syncytial Virus (RSV). Reduce secondary testing, decrease unnecessary antibiotic use and guide appropriate antiviral therapy.
  • Improve patient management with accurate, easy-to-read results in just 15 minutes
  • Eliminate workflow bottlenecks with less than one minute hands-on time and a simple, walk-away procedure
  • Stock up with 18-month shelf life at room-temperature storage
  • Offers flexible specimen storage, collection, and transport options
Xpect RSV uses antibody-coated color particles to bind with target antigen in the sample. The color-antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
DescriptionXpect RSV Test
IncludesTest devices, extraction buffer (2 x 10mL), disposable extraction tubes, disposable pipettes
Quantity30 Tests
Sample TypeNasopharyngeal
TypeReagent
Detectable AnalytesRespiratory Syncytial Virus (RSV) Antigen
Unit SizeEach
Contents & Storage
15°C to 30°C
MBD-0070-Coronavirus Viral Infections-278x123
Sample – Identify – Test
Find your product mix for coronavirus viral transport and associated viral and bacterial infections
#CoronavirusCare
Learn More 
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection