Thermo Scientific™

Wash buffer 2 for MagJET Plant RNA Kit (concentrated)

Catalog number: R2561
Have Questions?
Thermo Scientific™

Wash buffer 2 for MagJET Plant RNA Kit (concentrated)

Catalog number: R2561
Have Questions?
Catalog Number
R2561
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R256150 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific MagJet Wash Buffer 2 (conc.) is a component of the MagJET Plant RNA DNA Kits (K2771/2772) and may be purchased separately. Provided in 50 mL aliquots.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Buffer
Wash Buffer
For Use With (Equipment)
MagJET Plant RNA Kit
Product Line
MagJET
Quantity
50 mL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text