Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

TopVision Agarose Tablets

Catalog number: R2801
Have Questions?
Thermo Scientific™

TopVision Agarose Tablets

Catalog number: R2801
Have Questions?
Catalog Number
R2801
Unit Size
200 tablets (0.5g each)
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1,000 tablets (0.5g each)
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R2801200 tablets (0.5g each)
-
R28021,000 tablets (0.5g each)
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific TopVision Agarose Tablets, low EEO (LE), are a multi-purpose agarose in tablet format delivered in a convenient blister pack. The agarose tablets are made of standard melting point agarose that yields high resolution, sharp DNA bands with high clarity and low background. Its optimized gel strength enhances ease of gel processing and handling. Manufactured using an innovative Organic Solvent-Free manufacturing process that is greener and more environmentally friendly.

Features
• Convenient tablet format—no messy weighing required
• Fast dissolving protocol
• 'Green'—manufactured with significantly reduced environmental impact, using an innovative organic solvent-free technology.
• Enhanced resolution and reduced background

Applications

• Ideal for routine DNA and RNA gel electrophoresis and blotting assays.

Environmentally responsible Green Solution
Unlike traditional agarose powder, which production relies heavily on the use of toxic organic solvents, TopVision Agarose Tablets are manufactured using an innovative process that completely eliminates dependency on organic solvents and significantly reduces environmental impact while creating a safer working environment.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Top Vision Agarose Tablet
Separation Range
100 to >30000 bp
Shelf Life
2 Years
Melting Point
88 °C
Product Line
TopVision
Quantity
200 Tablets (0.5 g each)
Validated Application
Gel Electrophoresis

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text