Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Description | Detectable Analytes | Quantity |
---|---|---|---|
R30160201 | Salmonella H (a) | Salmonella H Antigen | 2 mL |
R30161601 | Salmonella H (i) | Salmonella H Antigen | 2 mL |
R30160401 | Salmonella H © | Salmonella H Antigen | 2 mL |
R30161401 | Salmonella H (gq) | Salmonella H Antigen | 2 mL |
R30160901 | Salmonella H (enz15) | Salmonella H Antigen | 2 mL |
R30160301 | Salmonella H (b) | Salmonella H Antigen | 2 mL |
R30160501 | Salmonella H (d) | Salmonella H Antigen | 2 mL |
R30161301 | Salmonella H (gp) | Salmonella H Antigen | 2 mL |
R30162901 | Salmonella H (z29) | Salmonella H Antigen | 2 mL |
R30163101 | Salmonella H (z38) | Salmonella H Antigen | 2 mL |
R30163301 | Salmonella H (1, 2) | Salmonella H Antigen | 2 mL |
R30163401 | Salmonella H (1, 5) | Salmonella H Antigen | 2 mL |
R30163001 | Salmonella H (z36) | Salmonella H Antigen | 2 mL |
R30858501 | Salmonella H Polyvalent, Phases 1 and 2 | Salmonella H Antigen | 2 mL |
R30162201 | Salmonella H™ | Salmonella H Antigen | 2 mL |
R30161701 | Salmonella H (k) | Salmonella H Antigen | 2 mL |
R30161501 | Salmonella H (gst) | Salmonella H Antigen | 2 mL |
R30161901 | Salmonella H (lv) | Salmonella H Antigen | 2 mL |
R30162601 | Salmonella H (z6) | Salmonella H Antigen | 2 mL |
R30162801 | Salmonella H (z27) | Salmonella H Antigen | 2 mL |
R30162101 | Salmonella H (mt) | Salmonella H Antigen | 2 mL |
R30160101 | Salmonella H Rapid Diagnostic 3 (d, E, G, k) | Salmonella H Antigen | 2 mL |
R30161801 | Salmonella H (L) Polyvalent, for lv, lw, lz13, lz28, lz38, lz40 | Salmonella H Antigen | 2 mL |
R30160601 | Salmonella H (E) Polyvalent, for eh, enx, enz15 | Salmonella H Antigen | 2 mL |
R30162501 | Salmonella H (z4, z23) | Salmonella H Antigen | 2 mL |
R30162401 | Salmonella H (z) | Salmonella H Antigen | 2 mL |
R30163201 | Salmonella H Polyvalent, Phase 2 (1, 2, 5, 6, 7) | Salmonella H Antigen | 2 mL |
R30959801 | Salmonella H Rapid Diagnostic 1 (b, d, E, r) | Salmonella H Antigen | 2 mL |
R30162701 | Salmonella H (z10) | Salmonella H Antigen | 2 mL |
R30959901 | Salmonella H Rapid Diagnostic 2 (b, E, k, L) | Salmonella H Antigen | 2 mL |
R30162001 | Salmonella H (lw) | Salmonella H Antigen | 2 mL |