Remel™ PYR Disc with Reagent
Remel™ PYR Disc with Reagent
Thermo Scientific™
Remel™ PYR Disc with Reagent
Use for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
R30854401100 Tests
R3085430150 Test
R21117225 Tests/Kit
Catalog number R30854401
Price (USD)
-
Request A Quote
Quantity:
100 Tests
Thermo Scientific™ Remel PYR Disk w/Reagent is used for presumptive identification of enterococci, Lancefield group A streptococci, and E. coli based on PYRase production.
Specifications
Quantity100 Tests
CE MarkerYes
Unit SizeEach
Contents & Storage
2°C to 8°C
Intended use statement
A kit used for the presumptive identification of Group A Streptococcus, enterococci and Escherichia coli by detection of pyrrolidonyl arylamidase (PYRase).
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link