Staphaurex™ Latex Agglutination Test
Staphaurex™ Latex Agglutination Test
Actual product may vary
Staphaurex™ Latex Agglutination Test
Staphaurex™ Latex Agglutination Test
Thermo Scientific™
Staphaurex™ Latex Agglutination Test
Rapidly identify and differentiate staphylococci which possess coagulase and/or protein A, particularly Staphylococcus aureus, from other staphylococci.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionQuantityContent And Storage
R30859901Staphaurex Latex Agglutination Test120 Tests/Kit
R30859902Staphaurex Latex Agglutination Test400 Tests/Kit
Catalog number R30859901
Price (USD)
-
Request A Quote
Description:
Staphaurex Latex Agglutination Test
Quantity:
120 Tests/Kit
Ask our AI about this Product
Rapidly identify and differentiate staphylococci which possess coagulase and/or protein A, particularly Staphylococcus aureus, using the Thermo Scientific™ Staphaurex™ Latex Agglutination Test.
Experience quality and reliability with this rapid and accurate latex test for the identification and differentiation of staphylococcus aureus. White latex particles are coated with human fibrinogen for detection of clumping factor and specific IgG for detection of protein A.
When mixed on a slide with a suspension of S. aureus, reaction of clumping factor with the fibrinogen and/or protein A with the IgG causes rapid, strong agglutination of the latex particles.
Proven Performance
  • Rely on tried-and-true tests with more than 50 years of combined proven performance
  • Maintain confidence with reliable results that have greater than 99% sensitivity and specificity (according to data available in the package inserts)
Workflow Efficiency
  • Clear-cut, robust reactions provide easy-to-read results
  • Improve productivity with compact kit configurations, color-coded reagents, and convenient workstations
  • Flexible use from selective and non-selective media
  • Control latex is not required
Economical Solution
  • Maximize spending efficiency with the option of purchasing separate kit components
  • Save time, money, and resources by turning to one company, one source, for a complete set of Staph identification solutions
Although Staphylococci are commonly found on the skin and in mucous membranes, they have been associated with many human and animal infections. S.aureus and other coagulase-positive Staphylococci have been identified as a cause of suppurative infections, food poisoning and toxic shock syndrome and have been isolated from nearly all anatomical sites.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Control SetsPositive
DescriptionStaphaurex Latex Agglutination Test
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity120 Tests/Kit
Target Organism ClassStaphylococcus aureus
TypeStaphylococcus aureus Test Kit
CE MarkerYes
Detectable AnalytesProtein A
Unit SizeEach
Contents & Storage
2°C to 8°C
Intended use statement
A qualitative latex slide agglutination test for the differentiation of Staphylococcus aureus from other Staphylococcus species isolates grown on agar by the detection of clumping factor and Protein A. Used in a diagnostic workflow to aid clinicians in treatment options for patients suspected of having bacterial infections. The device is not automated, is for professional use only and is not a companion diagnostic.
microbial-detection-system
Related product
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link