Thermo Scientific™

Staphaurex™ Plus Latex Agglutination Test

Catalog number: R30950102
Have Questions?
Thermo Scientific™

Staphaurex™ Plus Latex Agglutination Test

Catalog number: R30950102
Have Questions?
Identify staphylococci which possess clumping factor, protein A, and/or surface antigens, including those characteristic of Methicillin Resistant Staphylococcus aureus (MRSA) and Methicillin Sensitive Staphylococcus aureus (MSSA) strains, using the Thermo Scientific™ Staphaurex™ Plus Latex Agglutination Test.
 
Catalog Number
R30950102
Unit Size
Each
Description
Staphaurex Plus Latex Agglutination Test
Quantity
150 Tests/Kit
Content And Storage
Price (USD)
Full specifications
DescriptionStaphaurex Plus Latex Agglutination Test
CE MarkerYes
Detectable AnalytesStaphylococcus species
TypeStaphylococcus aureus Test Kit
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity150 Tests/Kit
Target Organism ClassStaphylococcus aureus
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Staphaurex Plus Latex Agglutination Test
Quantity
450 Tests/Kit
Content And Storage
Price (USD)
Full specifications
DescriptionStaphaurex Plus Latex Agglutination Test
CE MarkerYes
Detectable AnalytesStaphylococcus species
TypeStaphylococcus aureus Test Kit
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity450 Tests/Kit
Target Organism ClassStaphylococcus aureus
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionQuantityContent And StoragePrice (USD)
R30950102Full specifications
EachStaphaurex Plus Latex Agglutination Test150 Tests/KitRequest A Quote
DescriptionStaphaurex Plus Latex Agglutination Test
CE MarkerYes
Detectable AnalytesStaphylococcus species
TypeStaphylococcus aureus Test Kit
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity150 Tests/Kit
Target Organism ClassStaphylococcus aureus
Unit SizeEach
R30950201Full specifications
EachStaphaurex Plus Latex Agglutination Test450 Tests/KitRequest A Quote
DescriptionStaphaurex Plus Latex Agglutination Test
CE MarkerYes
Detectable AnalytesStaphylococcus species
TypeStaphylococcus aureus Test Kit
IncludesTest latex, disposable reaction cards, disposable sampling and mixing sticks
Quantity450 Tests/Kit
Target Organism ClassStaphylococcus aureus
Unit SizeEach
Showing 2 of 2
Use this rapid latex agglutination test which utilises yellow latex particles coated with human fibrinogen for the detection of clumping factor, specific IgG for the detection of protein A, and surface antigens characteristic of MRSA and MSSA strains.

In the presence of target organisms, the latex particles agglutinate to give a strong and easily visible positive reaction. In the absence of target organisms the particles remain in smooth suspension.

Proven Performance
  • Rely on tried-and-true tests with more than 50 years of combined proven performance
  • Maintain confidence with greater than 99% sensitivity (according to data available in the package inserts)

Workflow Efficiency
  • Clear-cut, robust reactions provide easy-to-read results
  • Improve productivity with compact kit configurations, color-coded reagents, and convenient workstations

Economical Solution
  • Maximize spending efficiency with the option of purchasing separate kit components
  • Save time, money, and resources by turning to one company, one source, for a complete set of Staph identification solutions

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Contents & Storage

2°C to 8°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text