Thermo Scientific™
Streptex™ Rapid Latex Agglutination Test
Catalog number: R30950555
Have Questions?
Thermo Scientific™
Streptex™ Rapid Latex Agglutination Test
Catalog number: R30950555
Have Questions?
Streptex™ Rapid is a rapid latex test system for use in the qualitative detection and identification of the Lancefield group of streptococci. Reagents are provided for groups A, B, C, F and G.
Catalog Number
R30950555
Unit Size
Each
Description
Streptex Rapid Latex Agglutination Test
Quantity
50 Tests/Kit
Price (USD)
Full specifications
Control SetsPositive
DescriptionStreptex Rapid Latex Agglutination Test
CE MarkerYes
Detectable AnalytesStreptococci (Group A, B, C, F, G)
TypeStreptococcus Test Kit
IncludesTest latex reagents, extraction reagents, polyvalent positive control, disposable tubes, disposable tube holder, disposable reaction cards, mixing sticks and sample dispensers
MaterialPolyvalent
Organism GroupLancefield A, B, C, F and G
Quantity50 Tests/Kit
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionQuantityPrice (USD)
R30950555Full specifications
EachStreptex Rapid Latex Agglutination Test50 Tests/KitRequest A Quote
Control SetsPositive
DescriptionStreptex Rapid Latex Agglutination Test
CE MarkerYes
Detectable AnalytesStreptococci (Group A, B, C, F, G)
TypeStreptococcus Test Kit
IncludesTest latex reagents, extraction reagents, polyvalent positive control, disposable tubes, disposable tube holder, disposable reaction cards, mixing sticks and sample dispensers
MaterialPolyvalent
Organism GroupLancefield A, B, C, F and G
Quantity50 Tests/Kit
Unit SizeEach
Showing 1 of 1
Use this rapid latex agglutination test which provides a complete solution for the isolation and differentiation of Lancefield groups A, B, C, F & G. View distinct, clear results with white latex agglutination on a black background. Individual latex grouping reagents are available as required.
With Streptex, group specific antigens are extracted from streptococci in a simple incubation step. Antigens are then identified using polystyrene latex particles which have been coated with group-specific antibodies. These latex particles agglutinate strongly in the presence of homologous antigen and remain in smooth suspension in the absence of homologous antigen.
For a more rapid acid extraction method, Streptex Acid Extraction Kit (Part No. R30951301) is available for use with Streptex. Acid extraction will rapidly extract the Group antigens without the need for incubation. However, Group D antigen is not extracted efficiently with acid extraction; therefore, an alternative method must be used to confirm suspected Group D streptococci.
Proven Performance
  • Rely on tried-and-true tests with more than 50 years of combined proven performance
  • Maintain confidence with reliable results that have greater than 99% sensitivity (according to data available in the package insert)
Workflow Efficiency
  • Get easy-to-read results with clear-cut, robust reactions
  • Improve productivity with compact kit configurations, color-coded reagents and convenient workstations
  • Enjoy flexibility with use from selective and non-selective media—no control latex required
Economical Solution
  • Maximize spending efficiency with the option of purchasing separate kit components
  • Save time, money and resources by turning to one company, one source, for a complete set of Staph identification solutions
Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma, according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Contents & Storage
2°C to 8°C