28mm Caps for Sterilin™ UC/30 Disposable Glass Containers
Product Image
Thermo Scientific™
28mm Caps for Sterilin™ UC/30 Disposable Glass Containers
Leak-free polypropylene caps for use with Thermo Scientific Sterilin Disposable Glass Containers.
Have Questions?
Catalog NumberNo. per Case
R3282850/Cs.
Catalog number R328
Price (USD)
-
Request A Quote
No. per Case:
2850/Cs.
Ask our AI about this Product
Ideal for small volume samples, Thermo Scientific™ Sterilin™ Disposable Glass Containers are made from soda glass with a leakfree polypropylene cap.
NOT AVAILABLE IN AMERICA
Specifications
Closure MaterialPolypropylene
Closure Size28 mm
Description28mm Caps for UC/30
For Use With (Equipment)39404 and 39503T Sterilin Disposable Glass Containers
No. per Case2850/Cs.
SterilityNon-sterile
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link
1x1 image pixel for data collection