Invitrogen™

NucRed™ Live 647 ReadyProbes™ Reagent

Catalog number: R37106
Have Questions?
Invitrogen™

NucRed™ Live 647 ReadyProbes™ Reagent

Catalog number: R37106
Have Questions?
Catalog Number
R37106
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R371061 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
NucRed Live 647 ReadyProbes Reagent is a bright, far-red, cell-permeant nuclear stain for live or fixed cells. It is provided as a room temperature-stable solution in a convenient-to-use dropper bottle. Just add two drops per ml to stain your cells.

Provided at 12.5X concentration and formulated with 1% DMSO, PowerLoad solubilizing agent, and HBSS buffer. Also available as HCS Nuclear Mask Deep Red Stain at 250X concentration in DMSO.

• No need to dilute, weigh, or pipette
• Convenient dropper bottle—just use two drops per mL
• Stable at room temperature—keep handy at your scope or cell culture area
• Excited at 638 nm when bound to DNA, with an emission maximum at 686 nm

See other ReadyProbes reagents for cell staining
See other nuclear stains for imaging

Cell imaging applications
The far-red fluorescence emission makes it ideal for use in combination with blue, green, and red fluorophores, such as Hoechst, GFP/FITC, and Texas Red.

Suggestions for use
• NucRed Live 647 ReadyProbes Reagent may be added directly to cells in full media, or buffer solutions.
• In most cases, 2 drops/mL and an incubation of 15 to 30 minutes will give bright nuclear staining; however, more or fewer drops can be added for optimal staining intensity.
• In most cases, staining intensity increases with time if cells are not washed prior to imaging.
• It is detected through a far-red filter, such as a Cy5 filter, or a Cy5 Light Cube for EVOS imaging systems.


Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Far-Red
Excitation Wavelength Range
638⁄686
Dye Type
Cell-Permeant
For Use With (Equipment)
Confocal Microscope, Fluorescence Microscope, Flow Cytometer
Quantity
6 dropper bottle(s)
Detection Method
Fluorescent
Emission
Visible
Form
Liquid
Product Line
NucRed™, ReadyProbes™
Label Type
Fluorescent Dye
Product Type
Nucleic Acid Stain
SubCellular Localization
Nucleus

Contents & Storage

6 × 2.5 mL dropper bottles

Store at ≤ 25°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text