Invitrogen™

ActinGreen™ 488 ReadyProbes™ Reagent (AlexaFluor™ 488 phalloidin)

Catalog number: R37110
Have Questions?
Invitrogen™

ActinGreen™ 488 ReadyProbes™ Reagent (AlexaFluor™ 488 phalloidin)

Catalog number: R37110
Have Questions?
Catalog Number
R37110
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R371101 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
ActinGreen 488 ReadyProbes Reagent is a selective, high-affinity F-actin probe (phalloidin) conjugated to our bright, photostable, green-fluorescent Alexa Fluor 488 dye. ActinGreen 488 reagent is formulated as a room temperature-stable solution of Alexa Fluor 488 Phalloidin (Cat. No. A12379) and comes in a convenient dropper bottle. Just tip and drip two drops per ml to stain your cells.

Also available: AlexaFluor 488 phalloidin at 12.5X concentration (66 U/mL) in PBS with BSA and trace amounts of methanol and sodium azide.

• High-affinity staining of F-actin with superior specificity compared to antibody methods
• Outstanding fluorescence performance (Ex/Em: 495/518 nm)
• Ready-to-use liquid formulation in convenient dropper bottle—no need to dilute, weigh, or pipette
• Stability at room temperature—keep handy at your scope or cell culture area

See other ReadyProbes reagents for cell staining
Learn more about other stains for actin

Cell imaging applications
Phalloidin is a bi-cyclic peptide, commonly used in imaging applications to selectively label F-actin. Fluorescently-labeled phalloidin has several advantages over antibodies for actin labeling, including virtually identical binding properties with actin from different species of plants and animals, and low non-specific binding. Phalloidin binds F-actin with high selectivity, while Alexa Fluor 488 dye provides green fluorescence of unparalleled brightness and photostability.

Suggestions for use
• ActinGreen 488 reagent may be added directly to fixed cells in full media or buffer solutions.
• In most cases, 2 drops/ml and an incubation time of 15 to 30 minutes is sufficient for bright actin staining; however, optimization may be needed for some cell types, conditions, and applications. In such cases, simply add more or fewer drops until the optimal staining intensity is obtained.
• ActinGreen 488 dye is excited with a maximum at 495 nm and has an emission maximum at 518 nm. It is detected through standard GFP and FITC filters.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Color
Green
Excitation Wavelength Range
495⁄518
Dye Type
Alexa Fluor™ 488
For Use With (Equipment)
Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Flow Cytometer, Floid™ Cell Imaging System
Quantity
2 dropper bottle(s)
Detection Method
Fluorescent
Emission
Visible
Form
Liquid
Product Line
ActinGreen™, ReadyProbes™
Technique
Fluorescence Intensity
Label Type
Fluorescent Dye
Product Type
Phalloidin
SubCellular Localization
Cytoskeleton

Contents & Storage

2 × 2.5 mL dropper bottles

Store at ≤ 25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text