Invitrogen™

ReadyProbes™ Avidin/Biotin Blocking Solution (1X)

Catalog number: R37627
Have Questions?
Invitrogen™

ReadyProbes™ Avidin/Biotin Blocking Solution (1X)

Catalog number: R37627
Have Questions?
Catalog Number
R37627
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
R376271 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
ReadyProbes Avidin/Biotin Blocking Solution blocks endogenous biotin, biotin receptors, and avidin binding sites present in many types of tissue, reducing background signal in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) procedures. This two-component solution (an avidin-blocking solution and a biotin-blocking solution) is provided in a ready-to-use format—just add a few drops of the avidin component and incubate for 15 minutes, then wash and repeat with the biotin component. ReadyProbes Avidin/Biotin Blocking Solution is used after the blocking of cells/tissue with standard non-specific blocking solution (e.g., diluted serum), but before the primary antibody step.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Avidin/Biotin Blocking Solution
Quantity
1 Kit
Product Line
ReadyProbes™
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice

Contents & Storage

A kit containing 18 mL each ready-to-use (1X) solutions of avidin (component A) and biotin (component B) in two clear LDPE dropper bottles.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text